Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

bcl-x Mouse anti-Human, Clone: BCL2L1/1762, Novus Biologicals™

Mouse Monoclonal Antibody

$138.37 - $352.89

Specifications

Antigen bcl-x
Clone BCL2L1/1762
Concentration 0.2 mg/ml
Dilution ELISA 1-5 ug/ml
Applications ELISA
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122306
View Documents
Novus Biologicals
NBP30741620UG
20 μg
Each for $138.37
Add to cart
 
NB122305
View Documents
Novus Biologicals
NBP30741610UG
100 μg
Each for $352.89
Add to cart
 
Description

Description

bcl-x Monoclonal antibody specifically detects bcl-x in Human samples. It is validated for ELISA
Specifications

Specifications

bcl-x
0.2 mg/ml
ELISA
Purified
RUO
10 mM PBS with 0.05% BSA
Recombinant human bcl-x protein fragment (around aa 1-135) (exact sequence is proprietary)
Monoclonal
Protein A or G purified
BCL2L1/1762
ELISA 1-5 ug/ml
Unconjugated
Mouse
Human
598
IgG2b Kappa
Primary
Store at 4°C.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit