Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
bcl-x Mouse anti-Human, Clone: SPM519, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP30741720UG
Description
bcl-x Monoclonal specifically detects bcl-x in Human samples. It is validated for Immunohistochemistry-Paraffin.Specifications
bcl-x | |
Monoclonal | |
0.2 mg/ml | |
Immunohistochemistry-Paraffin 1-2 ug/ml | |
Mouse | |
Protein A or G purified | |
RUO | |
598 | |
Store at 4°C. | |
IgG |
Immunohistochemistry (Paraffin) | |
SPM519 | |
Unconjugated | |
10 mM PBS with 0.05% BSA | |
Recombinant human bcl-x protein fragment (around aa 1-135) (exact sequence is proprietary) | |
20 μg | |
Primary | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title