Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

bcl-x Mouse anti-Human, Clone: SPM519, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

$265.00

Specifications

Antigen bcl-x
Clone SPM519
Concentration 0.2 mg/ml
Dilution Immunohistochemistry-Paraffin 1-2 ug/ml
Applications Immunohistochemistry (Paraffin)
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122308
SDP
View Documents
Novus Biologicals
NBP30741720UG
20 μg
Each of 1 for $265.00
Add to Cart
 
Description

Description

bcl-x Monoclonal specifically detects bcl-x in Human samples. It is validated for Immunohistochemistry-Paraffin.
Specifications

Specifications

bcl-x
0.2 mg/ml
Immunohistochemistry (Paraffin)
Unconjugated
Mouse
Human
598
IgG
Protein A or G purified
SPM519
Immunohistochemistry-Paraffin 1-2 ug/ml
Monoclonal
Purified
RUO
10 mM PBS with 0.05% BSA
Recombinant human bcl-x protein fragment (around aa 1-135) (exact sequence is proprietary)
Primary
Store at 4°C.
SDS
Documents

Documents

Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit