Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

bcl-x Mouse anti-Human, Clone: SPM519, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP30741710UG

 View more versions of this product

Catalog No. NB122307


Add to cart

Description

Description

bcl-x Monoclonal specifically detects bcl-x in Human samples. It is validated for Immunohistochemistry-Paraffin.
Specifications

Specifications

bcl-x
SPM519
Unconjugated
10 mM PBS with 0.05% BSA
Mouse
IgG
100 μg
Store at 4°C.
Monoclonal
Human
Immunohistochemistry (Paraffin)
0.2 mg/ml
Immunohistochemistry-Paraffin 1-2 ug/ml
Purified
Recombinant human bcl-x protein fragment (around aa 1-135) (exact sequence is proprietary)
Protein A or G purified
RUO
Primary
598
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit