Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bcl-xL Mouse anti-Human, Biotin, Clone: OTI4A9, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270247B

Catalog No. NBP270247B


Add to cart

Description

Description

Bcl-xL Monoclonal antibody specifically detects Bcl-xL in Human samples. It is validated for Western Blot, Immunocytochemistry, Immunofluorescence.
Specifications

Specifications

Bcl-xL
OTI4A9
Western Blot, Immunocytochemistry/Immunofluorescence
Affinity Purified, Conjugated
BCL2L1
Recombinant fragment expressed in E.coli corresponding to amino acids 1-212 of human Bcl-xL.
Immunogen affinity purified
Apoptosis, Biologically Active Proteins, Cancer, Phospho Specific, Tumor Suppressors
Primary
598.0
Immunocytochemistry, Immunofluorescence, Western Blot
Biotin
PBS with 0.05% Sodium Azide
Apoptosis regulator Bcl-X, bcl2-L-1, BCL2-like 1, bcl-2-like protein 1, BCLXBCL2LBcl-X, bcl-xL, BCLXL, BCL-XL/S, bcl-xS, BCLXS, DKFZp781P2092
Mouse
IgG2a
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit