Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BCL10 Mouse anti-Human, Clone: 151, Abnova™

Mouse monoclonal antibody raised against recombinant human BCL10.

Manufacturer:  Abnova Corporation MAB12881

Catalog No. 89-933-626


Add to cart

Description

Description

Specifications

Specifications

BCL10
151
Mouse monoclonal antibody raised against recombinant human BCL10.
In PBS (0.09% sodium azide).
CARMEN/CIPER/CLAP/c-E10/mE10
Mouse
IgG1
50 ug
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
ELISA, Flow Cytometry, Immunohistochemistry (PFA fixed), Immunoprecipitation, Western Blot
Unconjugated
ELISA Flow Cytometry Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (10 ug/mL) Immunoprecipitation Western Blot The optimal working dilution should be determined by the end user.
Liquid
BCL10
Recombinant protein corresponding to full length human BCL10.
Protein G purification
RUO
Primary
8915
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit