Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BCL10 Rabbit anti-Human, Unconjugated, Clone: BL10/2988R, Abnova™

Rabbit Monoclonal Antibody.

Manufacturer:  Abnova Corporation RAB00662

Catalog No. 89-339-677


Add to cart

Description

Description

BCL10 Monoclonal antibody specifically detects BCL10 in Human samples. It is validated for IHC-P, IF, Flow Cyt.
Specifications

Specifications

BCL10
BL10/2988R
B-cell CLL/lymphoma 10
Liquid
Rabbit
IgG
Store at -20 to -80°C
Monoclonal
Human
Immunohistochemistry (Paraffin), Immunofluorescence, Flow Cytometry
Unconjugated
BCL10
CARMEN, CIPER, CLAP, c-E10, mE10
Recombinant protein corresponding to amino acids 122-168 of human BCL10
100 μg
Primary
8915
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit