Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BCL2 Rabbit anti-Human, Unconjugated, Clone: BCL2/1878R, Abnova™

Rabbit Monoclonal Antibody.

Manufacturer:  Abnova Corporation RAB00618

Catalog No. 89-339-597


Add to cart

Description

Description

BCL2 Monoclonal antibody specifically detects BCL2 in Human samples. It is validated for WB-Ce, IHC-P.
Specifications

Specifications

BCL2
BCL2/1878R
B-cell CLL/lymphoma 2
Liquid
Rabbit
IgG
Store at -20 to -80°C
Monoclonal
Human
Western Blot, Immunohistochemistry (Paraffin)
Unconjugated
BCL2
Bcl-2
Recombinant protein corresponding to full-length human BCL2
100 μg
Primary
596
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit