Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BCL2L1 Mouse anti-Human, Clone: OTI2D1, liquid, TrueMAB
Mouse Monoclonal Antibody
Manufacturer: OriGene TA506188
Description
The protein encoded the BCL-X gene belongs to the BCL-2 protein family. BCL-2 family members form hetero- or homodimers and act as anti- or pro-apoptotic regulators that are involved in a wide variety of cellular activities. The proteins encoded by this gene are located at the outer mitochondrial membrane, and have been shown to regulate outer mitochondrial membrane channel (VDAC) opening. VDAC regulates mitochondrial membrane potential, and thus controls the production of reactive oxygen species and release of cytochrome C by mitochondria, both of which are the potent inducers of cell apoptosis. Alternative splicing results in multiple transcript variants encoding two different isoforms. The longer isoform acts as an apoptotic inhibitor and the shorter isoform acts as an apoptotic activator.Specifications
BCL2L1 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q07817 | |
BCL-XL/S, BCL2L, BCLX, Bcl-X, PPP1R52 | |
Mouse | |
IgG1 | |
100 μL | |
-20° C, Avoid Freeze/Thaw Cycles | |
Primary | |
598 |
Immunohistochemistry (Paraffin), Western Blot | |
OTI2D1 | |
Unconjugated | |
BCL2L1 | |
Liquid | |
BCL2L1 | |
Full length human recombit protein of human BCL2L1 produced in HEK293T cell. | |
Affinity Chromatography | |
RUO | |
Antibody | |
Monoclonal | |
Human |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title