Learn More
BCL2L12 Rabbit anti-Human, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 219691AP
Catalog No. 50-173-3662
Add to cart
Specifications
Specifications
BCL2L12
0.39 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9HB09
BCL2L12, BPR
Rabbit
IgG
150 μL
-20°C
Primary
83596
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
BCL2L12
Liquid
BCL2L12
BCL2L12 Fusion Protein Ag16856
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human