Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ BCL2L14 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5103612
Description
Antibody detects endogenous levels of total BCL2L14.
The protein encoded by this gene belongs to the BCL2 protein family. BCL2 family members form hetero- or homodimers and act as anti- or pro-apoptotic regulators that are involved in a wide variety of cellular activities. Overexpression of this gene has been shown to induce apoptosis in cells. Three alternatively spliced transcript variants encoding two distinct isoforms have been reported for this gene.
Specifications
BCL2L14 | |
Polyclonal | |
Unconjugated | |
BCL2L14 | |
4930452K23Rik; 4933405K19Rik; 9030625M01Rik; AI429211; Apoptosis facilitator Bcl-2-like protein 14; apoptosis regulator BCL-G; BCL2 like 14; Bcl2l14; bcl2-L-14; BCL2-like 14 (apoptosis facilitator); BCLG; testicular tissue protein Li 26 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
66813, 79370 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q9BZR8, Q9CPT0 | |
BCL2L14 | |
A synthesized peptide derived from human BCL2L14(Accession Q9BZR8), corresponding to amino acid residues T31-E81. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction