Sino Biological™ BCL6 Recombinant Human Protein, N-Trx.His tag Non-distribution product as customer accommodation.

Expressed from DNA sequence encoding human BCL6 N-terminal fragment (Met 1-Met 150) fused to Trx and polyhistidine tag at N-terminus

Manufacturer: Sino Biological™ 12083H30E50

 View more versions of this product

Catalog No. 12-083-H30E50

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


    Specifications

    Protein Tag Thioredoxin, His Tag
    Purity ≥90% by SDS-PAGE
    Source Human
    Common Name B-cell CLL/Lymphoma 6
    Expression System E.coli
    Product Type BCL6 Recombinant Human Protein
    Molecular Weight 34.2kd
    Quantity 50μg
    Structural Form Truncated, Recombinant
    Gene Symbol BCL6
    Validated Application Assay Standard
    Protein Family Transcription Factor
    For Use With (Application) Used as protein standard and in cell biology research applications
    Format Lyophilized
    Gene ID (Entrez) 604

    Expressed from DNA sequence encoding human BCL6 N-terminal fragment (Met 1-Met 150) fused to Trx and polyhistidine tag at N-terminus

    • Formulation: 20mM Tris, 10% glycerol, pH 8.0

  • Description & Specifications

Related Products


Product Content Feedback

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit