Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sino Biological™ BCL6 Recombinant Human Protein, N-Trx.His tag Non-distribution product as customer accommodation.

Expressed from DNA sequence encoding human BCL6 N-terminal fragment (Met 1-Met 150) fused to Trx and polyhistidine tag at N-terminus

Manufacturer:  Sino Biological™ 12083H30E25

 View more versions of this product

Catalog No. 12-083-H30E25


  Check Availability
Add to cart

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Specifications

Description

Expressed from DNA sequence encoding human BCL6 N-terminal fragment (Met 1-Met 150) fused to Trx and polyhistidine tag at N-terminus

  • Formulation: 20mM Tris, 10% glycerol, pH 8.0
Description & Specifications

Specifications

Used as protein standard and in cell biology research applications
604
≥90% by SDS-PAGE
Human
B-cell CLL/Lymphoma 6
Assay Standard
E.coli
Thioredoxin, His Tag
Lyophilized
34.2kd
5 x 5μg
Truncated, Recombinant
BCL6
Transcription Factor
BCL6 Recombinant Human Protein
Provide Content Correction

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit