Learn More
BCL7A Mouse anti-Human, Alexa Fluor™ 647, Clone: 15C, BD Pharmingen™
Mouse Monoclonal Antibody
Manufacturer:  BD Biosciences 566425
Catalog No. BDB566425
Add to cart
Description
Description
BCL7A Monoclonal antibody specifically detects BCL7A in Human samples. It is validated for Flow Cytometry,Immunofluorescence,Immunohistochemistry (Paraffin).
Specifications
Specifications
BCL7A
15C
Alexa Fluor 647
Mouse
50 μg
Refrigerate (2 to 8°C)
Monoclonal
Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin)
0.2 mg/mL
Purified
IgG1, κ
RUO
Primary
Human