Learn More
BCL7B Mouse anti-Human, Clone: OTI8E10, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF809576
Catalog No. 50-166-9597
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Positive regulator of apoptosis. Plays a role in the Wnt signaling pathway, negatively regulating the expression of Wnt signaling components CTNNB1 and HMGA1. Involved in cell cycle progression, maintece of the nuclear structure and stem cell differentiation. May play a role in lung tumor development or progression.
Specifications
Specifications
BCL7B
Monoclonal
Unconjugated
BCL7B
Lyophilized
BCL7B
Full length human recombit protein of human BCL7B produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Western Blot
OTI8E10
PBS with 8% trehalose and no preservative; pH 7.3
Q9BQE9
BCL tumor suppressor 7B
Mouse
IgG1
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
9275