Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BCL7B Mouse anti-Human, Clone: OTI2D9, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA527163

Catalog No. PIMA527163


Add to cart

Description

Description

Positive regulator of apoptosis. Plays a role in the Wnt signaling pathway, negatively regulating the expression of Wnt signaling components CTNNB1 and HMGA1 (PubMed:25569233). Involved in cell cycle progression, maintece of the nuclear structure and stem cell differentiation (PubMed:25569233). May play a role in lung tumor development or progression.
Specifications

Specifications

BCL7B
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q9BQE9
B-cell CLL/lymphoma 7 protein family member B; B-cell CLL/lymphoma 7B
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
9275
Western Blot
OTI2D9
Unconjugated
BCL7B
Liquid
BCL7B
Full length human recombit protein of BCL7B produced in E.coli
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit