Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BCL9L Mouse anti-Human, Clone:3B9C1, Abnova™

Mouse monoclonal antibody raised against recombinant human BCL9L.

Manufacturer:  Abnova Corporation MAB17573

Catalog No. 89-334-039


Add to cart

Specifications

Specifications

BCL9L
3B9C1
B-cell CLL/lymphoma 9-like
In PBS (0.05% sodium azide)
Liquid
Mouse
IgG1
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
ELISA, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
ELISA (1:10000)
Western Blot (1:500-1:2000)
Immunocytochemistry
Flow Cytometry
Immunohistochemistry (1:200-1:1000)
The optimal working dilution should be determined by the end user.
BCL9L
BCL9-2, DLNB11
Recombinant protein corresponding to amino acid 606-751 of human BCL9L from E. coli.
100μg
Antibody
283149
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit