Learn More
BCMA/TNFRSF17 Mouse anti-Human, Alexa Fluor™ 750, Clone: 1004023, R&D Systems™
Mouse Monoclonal Antibody
Manufacturer:  R&D Systems FAB1931S100UG
Catalog No. FB1931S100U
Add to cart
This item is not returnable. View return policy
Description
Description
BCMA/TNFRSF17 Monoclonal antibody specifically detects BCMA/TNFRSF17 in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
BCMA/TNFRSF17
1004023
Flow Cytometry 0.25-1 ug/10^6 cells
Q02223
B cell maturation antigen, B-cell maturation protein, BCMAtumor necrosis factor receptor superfamily member 17, BCMB-cell maturation factor, CD269, CD269 antigen, TNFRSF13A, TNFRSF17, tumor necrosis factor receptor superfamily, member 17
Mouse myeloma cell line NS0-derived recombinant human BCMA/TNFRSF17, Met1-Ala54, Accession No. Q02223
Protein A or G purified from hybridoma culture supernatant
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Monoclonal
Detects human BCMA/TNFRSF17 in direct ELISAs.
Flow Cytometry
Alexa Fluor™ 750
Supplied 0.2 mg/mL in a saline solution containing BSA and sodium azide.
Purified, Supernatant, Conjugated
Mouse
IgG2b
100 μg
Primary
608.0
Human