Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BCMA/TNFRSF17 Mouse anti-Human, Clone: 1004024, R&D Systems™
Mouse Monoclonal Antibody
$136.67 - $415.56
Specifications
Antigen | BCMA/TNFRSF17 |
---|---|
Clone | 1004024 |
Dilution | Flow Cytometry 0.25 ug/10^6 cells, ELISA, CyTOF-ready |
Applications | Flow Cytometry, ELISA, CyTOF |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB1932100
|
R&D Systems
MAB1932100 |
100 μg |
Each for $415.56
|
|
MAB1932SP
|
R&D Systems
MAB1932SP |
25 μg |
Each for $136.67
|
|
Description
BCMA/TNFRSF17 Monoclonal specifically detects BCMA/TNFRSF17 in Human samples. It is validated for Flow Cytometry, ELISA, CyTOF-ready.Specifications
BCMA/TNFRSF17 | |
Flow Cytometry 0.25 ug/10^6 cells, ELISA, CyTOF-ready | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
B cell maturation antigen, B-cell maturation protein, BCMAtumor necrosis factor receptor superfamily member 17, BCMB-cell maturation factor, CD269, CD269 antigen, TNFRSF13A, TNFRSF17, tumor necrosis factor receptor superfamily, member 17 | |
Mouse myeloma cell line NS0-derived recombinant human BCMA/TNFRSF17, Met1-Ala54, Accession # Q02223 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. |
1004024 | |
Flow Cytometry, ELISA, CyTOF | |
Unconjugated | |
Mouse | |
Human | |
Q02223 | |
608 | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title