Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BCMA/TNFRSF17, Mouse anti-Human, DyLight 650, Clone: BCMA/2366, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP279898C

Catalog No. NB012104


Add to cart

Description

Description

BCMA/TNFRSF17 Monoclonal antibody specifically detects BCMA/TNFRSF17 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin).
Specifications

Specifications

BCMA/TNFRSF17
BCMA/2366
ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Conjugated, Purified
TNFRSF17
Recombinant human BCMA/TNFRSF17 protein fragment (around aa 78-184) (exact sequence is proprietary)
Protein A or G purified
RUO
Store at 4°C in the dark.
Monoclonal
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
DyLight 650
50mM Sodium Borate with 0.05% Sodium Azide
B cell maturation antigen, B-cell maturation protein, BCMAtumor necrosis factor receptor superfamily member 17, BCMB-cell maturation factor, CD269, CD269 antigen, tumor necrosis factor receptor superfamily, member 17
Mouse
IgG2c Kappa
0.1 mL
Biologically Active Proteins, Cytokine Research, Immunology
Primary
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit