Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BCMO1 Polyclonal Antibody
BCMO1 Polyclonal Antibody
Supplier: Thermo Scientific PA523974
Description
BCMO1 Polyclonal Antibody for Western Blot

Specifications
BCMO1 | |
Polyclonal | |
Unconjugated | |
BCO1 | |
BCDO; BCDO1; BCMO; Bcmo1; BCO; BCO1; Beta,beta-carotene 15,15'-dioxygenase; beta,beta-carotene 15,15'-monooxygenase; beta-carotene 15 15'-dioxygenase; beta-carotene 15 15-dioxygenase; beta-carotene 15, 15'-dioxygenase; beta-carotene 15, 15-dioxygenase; beta-carotene 15, 15'-dioxygenase 1; beta-carotene 15,15' dioxygenase; beta-carotene 15,15'-monooxygenase; beta-carotene 15,15'-monooxygenase 1; beta-carotene cleavage enzyme; beta-carotene dioxygenase 1; Beta-carotene oxygenase 1; beta-CD; betaCMOOX; carotenoid-15,15'-oxygenase; CMO1; Cmoi | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
53630 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.4 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
Q9HAY6 | |
BCO1 | |
KLH conjugated synthetic peptide between 263-292 amino acids from the central region of human BCMO1. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction