Learn More
BCoR Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A301673A
Description
The recommended shelf life for this product is 1 year from date of receipt.
Transcriptional corepressor. May specifically inhibit gene expression when recruited to promoter regions by sequence-specific DNA-binding proteins such as BCL6 and MLLT3. This repression may be mediated at least in part by histone deacetylase activities which can associate with this corepressor. Involved in the repression of TFAP2A; impairs binding of BCL6 and KDM2B to TFAP2A promoter regions. Via repression of TFAP2A acts as a negative regulator of osteo-dentiogenic capacity in adult stem cells; the function implies inhibition of methylation on histone H3 ′Lys-4′ (H3K4me3) and ′Lys-36′ (H3K36me2). {ECO:0000269; PubMed:10898795, ECO:0000269; PubMed:15004558, ECO:0000269; PubMed:18280243, ECO:0000269; PubMed:19578371, ECO:0000269; PubMed:23911289}.Specifications
| BCoR | |
| Polyclonal | |
| Unconjugated | |
| BCOR | |
| ANOP2, MAA2, MCOPS2, BCL-6 corepressor, BCL-6 interacting corepressor, BCL6 co-repressor | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 54880 | |
| 4° C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.2 mg/ml | |
| TBS with 0.1% BSA and 0.09% sodium azide | |
| Q6W2J9 | |
| BCOR | |
| Between 1705 and 1755 | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.