Learn More
Berkshire™ BCR™ Polyester Full-Finger Cleanroom Glove Liners
Continuous filament knitted polyester construction is thin yet durable, while the full-finger design provides total protection for those with severe glove sensitivities.
$13.83
Specifications
Color White
Disposable No
Finger Style Full
Hand Ambidextrous
Material Polyester
View More Specs
Products 2
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
19-037189
View Documents
Berkshire™
BGL3.20R
Regular
Pack of 10 for $13.83
Case of 20 PK for $277.00
Add to cart
18-999-2002
View Documents
Berkshire™
BGL3.20L
Large
Pack of 10 for $13.83
Case of 20 PK for $277.00
Add to cart
Description
Description
  • Reduces perspiration and contact dermatitis commonly associated with wearing latex, urethane, PVC, or nitrile gloves
  • Full-finger design provides total protection for those with severe glove sensitivities
  • Can be worn alone as inspection gloves to prevent fingerprints on critical surfaces
  • 100% continuous filament knitted polyester is thin, yet durable
  • Ambidextrous design can be used on either hand
  • Gloves can be repeatedly laundered for reuse
  • Suitable for use in ISO Class Cleanrooms
Specifications
Specifications
White
Full
Polyester
BGL3
No
Ambidextrous
Knit