Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BCRP (ABCG2) Rabbit anti-Human,Mouse, Unconjugated, Clone: 10K5, MilliporeSigma™

Rabbit Monoclonal Antibody

Specifications

Antigen BCRP (ABCG2)
Applications Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
ZRB121725MI
View Documents
MilliporeSigma
ZRB121725UL
25 μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
ZRB12174XMI
View Documents
MilliporeSigma
ZRB12174X25UL
4 x 25 μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

BCRP (ABCG2) Monoclonal antibody specifically detects BCRP (ABCG2) in Human,Mouse samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blotting.
Specifications

Specifications

BCRP (ABCG2)
Unconjugated
Rabbit
ATP-binding cassette sub-family G member 2;Breast cancer resistance protein;CDw338;Mitoxantrone resistance-associated protein;Placenta-specific ATP-binding cassette transporter;Urate exporter;CD338
His-tagged recombinant fragment corresponding to 126 amino acids from the N-terminal, cytoplasmic domain of human Breast cancer resistance protein (BCRP).
Primary
Recommend storage of lyophilized product at 2 to 8°C. Avoid repeated freeze-thaws.
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Purified
Human, Mouse
ABCG2, ABCP, BCRP, BCRP1, MXR
Monoclonal
Protein A Purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit