Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BCS1L Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Manufacturer: Proteintech Group Inc 101752AP150UL

Description
This gene encodes a homolog of the S. cerevisiae bcs1 protein which is involved in the assembly of complex III of the mitochondrial respiratory chain. The encoded protein does not contain a mitochondrial targeting sequence but experimental studies confirm that it is imported into mitochondria. Mutations in this gene are associated with mitochondrial complex III deficiency and the GRACILE syndrome. Five alternatively spliced transcripts encoding the same protein have been described.Specifications
BCS1L | |
Polyclonal | |
Unconjugated | |
BCS1L | |
BCS1L, BJS, FLNMS, GRACILE, h BCS, h BCS1, Hs.6719, Mitochondrial chaperone BCS1, PTD | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
301514, 617, 66821 | |
Antibody | |
IgG |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9CZP5, Q9Y276 | |
BCS1L | |
BCS1L Fusion Protein Ag0231 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Liquid |
Safety and Handling
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title