Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD 1047 dihydrobromide, Tocris Bioscience™

Click to view available options
Quantity:
10mg
50mg

Chemical Identifiers

138356-20-4
C13H20Cl2N2
275.217
MGVRNMUKTZOQOW-UHFFFAOYSA-N
N-[2-(3, 4-Dichlorophenyl)ethyl]-N-methyl-2-(dimethylamino)ethylamine dihydrobromide
188914

σ1 selective antagonist

σ1 receptor antagonist, exhibiting a similar binding profile to that of BD 1063 (Cat. No. 0883) but with higher affinity (approximately 10-fold) at both σ1 and σ2 sites.
Quantity 50mg
SMILES CN(C)CCN(C)CCC1=CC(=C(C=C1)Cl)Cl
IUPAC Name N'-[2-(3,4-dichlorophenyl)ethyl]-N,N,N'-trimethylethane-1,2-diamine
Formula Weight 437.04
Chemical Name or Material BD 1047 dihydrobromide
Grade >99%

Recommended Storage : Desiccate at +4°C