Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD 1063 dihydrochloride, Tocris Bioscience™ Available on GSA/VA Contract for Federal Government customers only.

Click to view available options
Quantity:
10mg
50mg

Chemical Identifiers

206996-13-6
C13H20Cl4N2
346.117
NXFDBTLQOARIMH-UHFFFAOYSA-N
1-[2-(3, 4-Dichlorophenyl)ethyl]-4-methylpiperazine dihydrochloride
11617161

Selective σ1 ligand, putative antagonist

σ1 receptor antagonist, approximately 50-fold selective over σ2 sites and ≥100-fold selective over opioid, PCP, muscarinic, dopamine, α1, α2, β adrenoceptor, 5-HT1 and 5-HT2 receptors. Prevents hyperlocomotion following cocaine administration in mice.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Quantity 10mg
SMILES CN1CCN(CC1)CCC2=CC(=C(C=C2)Cl)Cl.Cl.Cl
IUPAC Name 1-[2-(3,4-dichlorophenyl)ethyl]-4-methylpiperazine;dihydrochloride
Formula Weight 346.13
Chemical Name or Material BD 1063 dihydrochloride
Grade >99%

Recommended Storage : Store at Room Temperature