Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Akt/PKB Sampler Kit

For intracellular transduction of insulin-induced signals and anti-apoptotic effects of multiple survival factors

Manufacturer:  BD 611437

Catalog No. BDB611437


Add to cart

Includes: Bad, GSK-3b, Insulin Receptor b, IRS-1, p70s6k, PI3-Kinase, PKBa/Akt, PKB Kinase/PDK1, PP2A Catalytic a

Description

Description

  • Format: Purified
  • Storage buffer: Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide

Western Blotting

Specifications

Specifications

1 Kit
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

Research Use Only