Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BD BBL™ Brucella Agar with 5% Sheep Blood, Hemin and Vitamin K1 Prepared Media
For the isolation and cultivation of fastidious, obligately anaerobic organisms
$788.00
Specifications
Certifications/Compliance | ISO/DIS, 11014 |
---|---|
Color | Red |
Form | Liquid |
Manufacturing Origin | United States |
Formulation | Pancreatic digest of Casein 10g/L, Peptic digest of Animal Tissue 10g/L, Dextrose 1g/L, Yeast extract 2g/L, Sodium Chloride 5g/L, Sodium Bisulfite 0.1g/L, Agar 15g/L, Hemin 0.005g/L, Vitamin K1 0.01g/L, Sheep Blood, defibrinated 5% |
Catalog Number | Mfr. No. | dimMediaType | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | dimMediaType | Quantity | Price | Quantity & Availability | ||||
B4397716X
|
BD
297716 |
100/Pk. |
Pack of 100 for $788.00
|
|
|||||
L97848
|
BD
297848 |
20/Pk. | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
Description
Specifications
ISO/DIS, 11014 | |
Liquid | |
Pancreatic digest of Casein 10g/L, Peptic digest of Animal Tissue 10g/L, Dextrose 1g/L, Yeast extract 2g/L, Sodium Chloride 5g/L, Sodium Bisulfite 0.1g/L, Agar 15g/L, Hemin 0.005g/L, Vitamin K1 0.01g/L, Sheep Blood, defibrinated 5% | |
Plate |
Red | |
United States | |
Fastidious, obligately anaerobic organisms |
Safety and Handling
ShelfLife : 70 Days
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title