Learn More
BD BBL™ D/E Neutralizing Agar Sterile Pack Media - Rodac™ Plates
Plated media in gamma-irradiated packs for isolator systems—multiwrap packaging protects media from vaporized hydrogen peroxide exposure
Manufacturer:  BD 222209
 View more versions of this product
Catalog No. 08-757-186
Add to cart
This item is not returnable. View return policy
Description
Description
Rodac (Replicate Organism Detection and Counting) plates for surface sampling.
To use:
  • Place wrapped package of plates in the isolator before running the vaporized hydrogen peroxide decontamination cycle
  • When the isolator aeration cycle has finished, remove the two outermost wrappings, then the sterile inner wrapping
  • Use the included inner bag to transport plates from your critical environment
Compliance
Tested for performance and sterility according to Association for the Advancement of Medical Instrumentation (AAMI) guidelines.
Specifications
Specifications
ISO/DIS, 11014
Rodac
D/E neutralizing Agar (Rodac™)
100/Pk.
Sterile
Purple
United States
Plate
2-8°C
Safety and Handling
Safety and Handling
ShelfLife : 112 Days