Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Bacto™ Proteose Peptone Available on GSA/VA Contract for Federal Government customers only.

Used in preparing microbiological culture media and in producing bacterial toxins. Gibco™ Bacto™ Proteose Peptone is a highly nutritious, animal-based media. The enzymatic digest of protein is high in proteoses and provides nitrogen in a form that is readily available for bacterial growth.

$305.18 - $6079.13

Specifications

Format Dehydrated, free-flowing granules
For Use With (Application) Formulation of culture media for the cultivation of heterotrophic microorganisms
Shelf Life 1825 Days
Storage Requirements <30°C
pH 6.6–7.6 (1.0% solution at 25°C)
View More Specs

Products 2
Catalog Number Mfr. No. Packaging Quantity With Additives Price Quantity & Availability  
Catalog Number Mfr. No. Packaging Quantity With Additives Price Quantity & Availability  
DF0120-17-6 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Gibco™
211684
Bottle 500g Proteose Peptone
Each for $305.18
Add to cart
 
D012008
View Documents
Gibco™
212010
Pail 10kg Proteose Peptone
Each for $6,079.13
Add to cart
 
Description

Description

Proteose Peptone is an animal-based ingredient used in preparing microbiological culture media and in producing bacterial toxins. It may be used in culture medium for a variety of applications, including production of substances from the culture of bacteria, fungi, and mammalian cells. It has been reported that Proteose Peptone as supplementation to defined medium resulted in significant increases in cell number and specific monoclonal antibody production in batch culture systems.

Many factors account for the suitability of Bacto™ Proteose Peptone for the culture of fastidious pathogens, including the nitrogen components, buffering range, and the high content of proteoses. These elements create an environment beneficial to the maintenance of virulence and the elaboration of bacterial by-products. Thus, stock cultures are well preserved on media containing Bacto™ Proteose Peptone.

  • All animal tissues used are sourced in a controlled and validated way as to reduce risk of contamination or exposure to any diseases, especially Bovine Spongiform Encephalopathy (BSE).
  • Used in producing bacterial toxins, including: diphtheria toxin fom Corynebacterium diphtheria and toxins from Clostridium botulinum, Clostridium perfringens, hemolytic streptococci, pneumococcus, Salmonella pullorum (Salmonella cholerasuis subsp. cholerasuis).
  • This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Specifications

Dehydrated, free-flowing granules
1825 Days
6.6–7.6 (1.0% solution at 25°C)
Formulation of culture media for the cultivation of heterotrophic microorganisms
<30°C
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit