Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BD BBL™ Mycoflask™ Moeller Decarboxylase Broth with Lysine Prepared Media
Used for the biochemical differentiation of gram-negative enteric bacilli based on the production of lysine
Supplier: BD 221661
This item is not returnable.
View return policy
Description
Specifications
CE, SMD, SMWW, USDA | |
2°C to 8°C | |
United States | |
Peptic digest of Animal Tissue 5g/L, Beef extract 5g/L, Bromcresol Purple 0.01g/L, Cresol red 0.005g/L, Dextrose 0.5g/L, Pyridoxal 0.005g/L | |
Tube with Tight Cap |
Beige | |
Liquid | |
Moeller Decarboxylase Broth | |
Used for the biochemical differentiation of gram negative enteric bacilli | |
5 mL |
Safety and Handling
ShelfLife : 365 Days
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For in vitro Diagnostic Use.
Spot an opportunity for improvement?
Provide Content Correction