Learn More
BD BBL™ Mycoflask™ Motility Indole Ornithine (MIO) Deeps Prepared Media
To demonstrate motility, indole production and ornithine decarboxylase activity for the differentiation of Enterobacteriaceae
Manufacturer:  BD 221518
 View more versions of this product
Catalog No. L21518
Add to cart
This item is not returnable. View return policy
Description
Description
Specifications
Specifications
Pancreatic digest of Casein 9.5g/L, Pancreatic digest of Gelatin 10g/L, Yeast extract 3g/L, Dextrose 1.5g/L, L-Ornithine Monohydrochloride 5g/L, Bromcresol Purple 0.02g/L, Agar 2g/L
US
CE, BAM
Liquid
To demonstrate motility, indole production and Ornithine decarboxylase activity for the differentiation of Enterobacteriaceae
100 Pack
Tube
MIO Medium
Safety and Handling
Safety and Handling
ShelfLife : 365 Days
SDS
Documents
Product Certifications
Promotions
Promotions
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For in vitro Diagnostic Use.