Learn More
BD BBL™ Mycoflask™ Motility Indole Ornithine (MIO) Deeps Prepared Media
To demonstrate motility, indole production and ornithine decarboxylase activity for the differentiation of Enterobacteriaceae
$59.60 - $510.00
Specifications
Certifications/Compliance CE, BAM
Format Liquid
For Use With (Application) To demonstrate motility, indole production and Ornithine decarboxylase activity for the differentiation of Enterobacteriaceae
Manufacturing Origin US
View More Specs
Products 2
Catalog Number Mfr. No. Media Type Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Media Type Quantity Price Quantity & Availability  
BB21517
View Documents Promotion Details
BD
221517
MIO Medium 10 Pack
Pack of 10 for $59.60
Add to cart
L21518
View Documents Promotion Details
BD
221518
MIO Medium 100 Pack
Pack of 100 for $510.00
Add to cart
Description
Description
Specifications
Specifications
CE, BAM
Liquid
To demonstrate motility, indole production and Ornithine decarboxylase activity for the differentiation of Enterobacteriaceae
US
Safety and Handling
Safety and Handling
ShelfLife : 365 Days
SDS
Product Certifications
For in vitro Diagnostic Use.