Learn More
BD BBL™ Mycoflask™ Mueller Hinton II Broth (Cation-Adjusted) Prepared Media
For use in quantitative procedures for susceptibility testing of aerobic and facultatively anaerobic bacteria from clinical specimens
Manufacturer:  BD 297963
 View more versions of this product
Catalog No. L4397963
Add to cart
Description
Description
  • Formulated to have low thymine and thymidine content
  • Consisting of microorganisms, nutrient medium and antimicrobial agent
Specifications
Specifications
Light Beige
Susceptibility testing of rapidly growing aerobic and facultatively anaerobic bacteria isolated from clinical specimens
400 mL
Bottle
Mueller Hinton II Broth
Beef extract 3g/L, Acid Hydrolysate of Casein 17.5g/L, Starch 1.5g/L
US
ISO/DIS, 11014, BS12, CLSI, CMPH2, MCM9
Liquid
Safety and Handling
Safety and Handling
ShelfLife : 365 Days
SDS
Documents
Product Certifications
Promotions
Promotions
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For in vitro Diagnostic Use.