Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BD BBL™ Mycoflask™ Mycosel™ Agar Prepared Media
For the isolation of pathogenic fungi from materials having large amount of flora of other fungi and bacteria
Supplier: BD 221130
This item is not returnable.
View return policy
Description
- Contains cycloheximide and chloramphenicol
- For recovery of dermatophytes
Specifications
ISO/DIS, 11014, BS12, CMPH2, MCM9 | |
2°C to 8°C | |
United States | |
Isolation of pathogenic fungi, Contains cycloheximide/chloramphenicol | |
10/Pk. |
Light Yellow | |
Liquid | |
Papaic digest of Soybean Meal 10g/L, Dextrose 10g/L, Agar 15.5g/L, Cycloheximide 0.4g/L, Chloramphenicol 0.05g/L | |
Mycoflask™ Bottle |
Safety and Handling
ShelfLife : 274 Days
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For in vitro Diagnostic Use.
Spot an opportunity for improvement?
Provide Content Correction