Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BD BBL™ Mycoflask™ Sabouraud Dextrose Agar Prepared Media Slants
Used in qualitative procedures for cultivation of dermatophytes
$608.00
Specifications
Certifications/Compliance | CE, BAM, CCAM, CMPH2, COMPF, EP, JP, MCM9, USP |
---|---|
Color | Beige |
Content And Storage | 2°C to 8°C |
Form | Liquid |
Manufacturing Origin | United States |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
BB21013
|
BD
221013 |
100 Pack |
Pack of 100 for $608.00
|
|
|||||
BB21012
|
BD
221012 |
10/Pk. |
Item Discontinued
This item has been discontinued by the supplier. Please Sign In to view product availability in your area. View alternate product |
N/A | |||||
Description
Tubes with tight caps should be opened carefully to avoid injury due to breakage of glass.
Specifications
CE, BAM, CCAM, CMPH2, COMPF, EP, JP, MCM9, USP | |
2°C to 8°C | |
United States | |
Pancreatic digest of Casein 5g/L, Peptic digest of Animal Tissue 5g/L, Dextrose 40g/L, Agar 15g/L | |
Tube with Tight Cap |
Beige | |
Liquid | |
Sabouraud Dextrose Agar | |
Cultivation of dermatophytes |
Safety and Handling
ShelfLife : 548 Days
For in vitro diagnostic use.
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title