Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BD BBL™ Mycoflask™ Sabouraud Dextrose Agar Prepared Media Slants with Chloramphenicol
Used in qualitative procedures for cultivation of dermatophytes
Supplier: BD 221314
This item is not returnable.
View return policy
Description
Tubes with tight caps should be opened carefully to avoid injury due to breakage of glass.
Specifications
ISO/DIS, 11014, MCM9 | |
Liquid | |
Sabouraud Dextrose Agar | |
Cultivation of dermatophytes | |
10/Pk. |
2°C to 8°C | |
United States | |
Pancreatic digest of Casein 5g/L, Peptic digest of Animal Tissue 5g/L, Dextrose 40g/L, Agar 15g/L | |
Tube with Tight Cap |
Safety and Handling
ShelfLife : 365 Days
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For in vitro diagnostic use.
Spot an opportunity for improvement?
Provide Content Correction