Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BD BBL™ Mycoflask™ Trypticase™ Soy Agar Slants Prepared Media
Used for the isolation and cultivation of fastidious and nonfastidious microorganisms, including anaerobic and aerobic bacteria
$477.00
Specifications
Certifications/Compliance | ISO/DIS, 11014, AOAC, BAM, CCAM, COMPF, EP, EPA, ISO, JP, SMD, SMWW, USDA, USP |
---|---|
Color | Beige |
Form | Liquid |
Manufacturing Origin | United States |
Media Type | Trypticase Soy Agar |
Catalog Number | Mfr. No. | For Use With (Application) | Packaging Type | Quantity | Content And Storage | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | For Use With (Application) | Packaging Type | Quantity | Content And Storage | Price | Quantity & Availability | ||
BB21087
|
BD
221087 |
Presumptive identification of Helicobacter pylori in gastric antral biopsy specimens | Tube with Tight Cap | 100 Pack | 2 to 25°C |
Pack of 100 for $477.00
|
|
||
BB21086
|
BD
221086 |
Isolation and cultivation of fastidious as well as non-fastidious microorganisms, including anaerobic as well as aerobic bacteria, although it is not the medium of choice for anaerobes | Tube | 10/Pk. | 15°C to 30°C |
Item Discontinued
This item has been discontinued by the supplier. Please Sign In to view product availability in your area. View alternate product |
N/A | ||
Description
Tubes with tight caps should be opened carefully to avoid injury due to breakage of glass.
Specifications
ISO/DIS, 11014, AOAC, BAM, CCAM, COMPF, EP, EPA, ISO, JP, SMD, SMWW, USDA, USP | |
Liquid | |
Trypticase Soy Agar |
Beige | |
United States | |
Pancreatic digest of Casein 15g/L, Papaic digest of Soybean Meal 5g/L, Sodium Chloride 5g/L, Agar 15g/L |
Safety and Handling
ShelfLife : 548 Days
For in vitro diagnostic use.
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title