Learn More
BD BBL™ Prepared Plated Media: Mueller Hinton Chocolate Agar
For use in qualitative procedures for the isolation and cultivation of fastidious organisms, particularly Haemophilus species
Manufacturer:  BD 221869
 View more versions of this product
Catalog No. L21869
Add to cart
This item is not returnable. View return policy
Description
Description
The media is recommended by CLSI for antimicrobial susceptibility testing of bacteria isolated from animals.
  • Ready-to-use
  • 100 x 10mm, 100 x 15mm and 150 x 15mm-style plates
Specifications
Specifications
Dark Brown
Qualitative procedures for the isolation and cultivation of fastidious organisms, particularly Haemophilus species
8/Pk.
Plate
Mueller Hinton Chocolate Agar
Beef extract 2g/L, Acid Hydrolysate of Casein 17.5g/L, Starch 1.5g/L, Ag/Lar 17g/L, Hemog/Llobin 10g/L, IsoVitaleX™ enrichment (0.01g/L Vitamin B12 10mL, L-Glutamine 10g/L, Adenine 1g/L,g/Luanine Hydrochloride 0.03g/L, Aminobenzoic Acid
US
ISO/DIS, 11014
Liquid
Safety and Handling
Safety and Handling
ShelfLife : 119 Days