Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BD BBL™ Prepared Plated Media: Mueller Hinton Chocolate Agar
For use in qualitative procedures for the isolation and cultivation of fastidious organisms, particularly Haemophilus species
Specifications
Certifications/Compliance | ISO/DIS, 11014 |
---|---|
Color | Dark Brown |
Form | Liquid |
Manufacturing Origin | United States |
Media Type | Mueller Hinton Chocolate Agar |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
L21869
|
BD
221869 |
8/Pk. |
Item Discontinued
This item has been discontinued by the supplier. Please Sign In to view product availability in your area. View alternate product |
N/A | |||||
Description
The media is recommended by CLSI for antimicrobial susceptibility testing of bacteria isolated from animals.
- Ready-to-use
- 100 x 10mm, 100 x 15mm and 150 x 15mm-style plates
Specifications
ISO/DIS, 11014 | |
Liquid | |
Mueller Hinton Chocolate Agar | |
Plate |
Dark Brown | |
United States | |
Qualitative procedures for the isolation and cultivation of fastidious organisms, particularly Haemophilus species |
Safety and Handling
ShelfLife : 119 Days
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title