Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Prepared Plated Media: Sabouraud Dextrose Agar w/Chloramphenicol and Gentamicin (Deep Fill)

Used in qualitative procedures for cultivation of dermatophytes

Manufacturer:  BD 296359

Catalog No. L96359


Add to cart
This item is not returnable. View return policy

Description

Description

  • Ready-to-use
  • 100 x 10mm and 100 x 15mm-style plates
  • Deep-filled plates reduce the effects of drying during prolonged incubation

Specifications

Specifications

Light Beige
Cultivation of dermatophytes
20/Pk.
Plate
Sabouraud Dextrose agar with Chloramphenicol and Gentamicin
Pancreatic digest of Casein 5g/L, Peptic digest of Animal Tissue 5g/L, Dextrose 40g/L, Agar 15g/L, Chloramphenicol 0.05g/L,g/Lentamicin 0.05g/L
US
ISO/DIS, 11014
Liquid
Safety and Handling

Safety and Handling

ShelfLife : 147 Days

SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit