Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BD BBL™ Prepared Plated Media: Schaedler K-V Agar with 5% Sheep Blood
Highly nutritious, selective media for the isolation and cultivation of fastidious gram-negative anaerobic microorganisms, especially Bacteroides and Prevotella species
Supplier: BD 221555
This item is not returnable.
View return policy
Description
This media particularly is useful in the selective isolation of gram-negative anaerobes due to the inclusion of kanamycin and vancomycin in its formulation.
- Ready-to-use
- 100 x 10mm and 100 x 15mm-style plates
Specifications
ISO/DIS, 11014 | |
2°C to 8°C | |
United States | |
Pancreatic digest of Casein 8.2g/L, Peptic digest of Animal Tissue 2.5g/L, Papaic digest of Soybean Meal 1g/L, Dextrose 5.8g/L, Yeast extract 5g/L, Sodium Chloride 1.7g/L, Dipotassium Phosphate 0.8g/L, L-Cystine 0.4g/L, Hemin 0.01g/L, Vitamin K1 0.01g/L, | |
Plate |
Red | |
Solid | |
Schaedler K-V agar with 5% Sheep blood | |
Fastidious gram negative anaerobic microorganisms | |
20/Pk. |
Safety and Handling
ShelfLife : 56 Days
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction