Learn More
BD BBL™ Prepared Plated Potato Flakes Agarft. Available on GSA/VA Contract for Federal Government customers only.
Used in qualitative procedures for the cultivation of pathogenic and opportunistic fungi encountered in clinical mycology
$52.00 - $401.00
Specifications
Packaging Plate
Manufacturing Origin United States
Storage Requirements 2-8°C
View More Specs
Products 2
Catalog Number Mfr. No. Color Format Media Type Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Color Format Media Type Quantity Price Quantity & Availability  
L98328 Available on GSA/VA Contract for Federal Government customers only.
View Documents
BD
298328
Light Red Liquid Potato flakes Agar 10/Pk.
Pack of 10 for $52.00
Add to cart
B292362
View Documents
BD
292362
Light Beige Solid Potato flakes Agar with Chloramphenicol and Gentamicin 100/Pk.
Case of 100 for $401.00
Add to cart
Description
Description
  • Induces sporulation, enhancing the production of morphological structures required for identification
  • Addition of antimicrobial agents provides selectivity for more effective isolation and identification of medically significant fungi
  • Part No. 292362 includes Potato Flakes Agar with Chloramphenicol and Gentamicin, which helps to improve the recovery of pathogenic fungi from specimens heavily contaminated with bacteria and saprophytic fungi
  • Chloramphenicol inhibits a wide range of gram-positive and gram-negative bacteria, while Gentamicin is an aminoglycoside antibiotic that inhibits growth of gram-negative bacteria
For in vitro Diagnostic Use.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Plate
United States
2-8°C