Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BD BBL™ Prepared Sterile Pack Settling Plated Trypticase™ Soy Agar
Plated media in gamma-irradiated packs for isolator systems-multiwrap packaging protects media from vaporized hydrogen peroxide exposure
$1242.00 - $2003.00
Specifications
Certifications/Compliance | ISO/DIS, 11014 |
---|---|
Color | Light Beige |
Content And Storage | 2°C to 8°C |
Manufacturing Origin | United States |
Media Type | Trypticase™ Soy Agar |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
08-757-189
|
BD
222206 |
45/Pk. |
Pack of 45 for $2,003.00
|
|
|||||
08-757-188
|
BD
222205 |
100/Pk. |
Pack of 100 for $1,242.00
|
|
|||||
B21236X
|
BD
221236 |
10/Pk. | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
Description
- Available in 100 x 15 and 150 x 15mm style plates
Specifications
ISO/DIS, 11014 | |
2°C to 8°C | |
Trypticase™ Soy Agar | |
Plate |
Light Beige | |
United States | |
Sterile |
Safety and Handling
ShelfLife : 154 Days
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title