Learn More
BD Gram Decolorizer
Can be used as a replacement reagent for the BD Gram Stain Kits. BD Gram Decolorizer is used in a 4-step Gram staining process, before counterstaining. Available in 250ml solution bottles for direct use or gallon containers for bulk storage.
Manufacturer: BD 212528
Description
Gram Staining is used to differentiate gram-positive and gram-negative microorganisms from cultures or specimens. This Gram Decolorizer is an acetone (25%)/isopropanol (75%) solution for rapid decolorization, prior to counterstaining.
- Complete stain kits reduce the mess associated with Gram staining procedures
- Break-resistant solution bottle features a hinged cap that makes regulating the flow easy and helps prevent spots and spills
- Color-coded label allows quick identification of reagent
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Gram Decolorizer | |
CE | |
Staining microorganisms from cultures or specimens by the differential Gram method. | |
No drip flip cap | |
15° to 30°C | |
Decolorizer |
1 gallon / 3.8L | |
Liquid | |
Clear ad colorless | |
Acetone (25%), Isopropanol (75%) | |
Bulk storage container |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.