BD BBL™ RODAC™ SL Prepared Sterile Pack Plated Media
For use in cleanrooms and isolators
Manufacturer: BD 222243
 View all options for this product
Catalog No. 22-349-791
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »
    Format Rodac
    Media Type D/E Neutralizing Agar
    Quantity 10/Pk.
    Sterile Yes
    • RODAC (Replicate Organism Detection and Counting) prepared contact plates for environmental monitoring
    • Plate with secure lid and marked grid, loose fitting lid provides fast and easy one hand manipulation
    • Three lugs on base of plate provide tight fit between lid and base to reduce accidental contamination
    • Plates filled with different types of prepared media
    For use in air sampling equipment such as the Surface Air System
  • Description & Specifications
Related Products
Product Content Feedback