Learn More
BD BBL™ Sensi-Disc™ Antimicrobial Susceptibility Test Discs, Cefuroxime27
Used for semi-quantitative in vitro testing by agar disc diffusion test procedure of certain fastidious bacterial pathogens
Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
B31621
View Documents
BD
231621
10 Pack
Pack of 10 for $221.00
N/A
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
  • 6mm discs impregnated with accurately determined amounts of antibiotic or chemotherapeutic agents
  • Are clearly marked on both sides with letters and numbers designating agent and drug content
Safety and Handling
Safety and Handling
ShelfLife : 730 Days
SDS
Product Certifications
For in vitro diagnostic use