Learn More
BD BBL™ Sensi-Disc™ Susceptibility Test Discs: Fluconazole, triazole
High-quality absorbent paper discs impregnated with accurately determined amounts of antibiotic or other chemotherapeutic agents
Manufacturer:  BD 232201
Catalog No. 22-349-960
Add to cart
This item is not returnable. View return policy
Description
Description
  • Discs containing certifiable agents are submitted to FDA for assay and certification before distribution Clearly marked on both sides with letters and numbers designating agent and concentration
Specifications
Specifications
Fluconazole, triazole
Disc
Each
25μg
Safety and Handling
Safety and Handling
ShelfLife : 365 Days