Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Difco™ Tryptone Peptone

$175.00 - $3383.00

Specifications

Format Powder
For Use With (Application) Dehydrated culture media
Shelf Life 1460 Days
With Additives Peptone
View More Specs

Products 3
Catalog Number Mfr. No. Packaging Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Packaging Quantity Price Quantity & Availability  
B11921
Accessories View Documents
Gibco™
211921
Bottle 1 lb.
Each for $175.00
Add to cart
 
B11922
View Documents
Gibco™
211922
Pail 5 lb.
Each for $792.00
Add to cart
 
B11923
View Documents
Gibco™
211923
Pail 25 lb.
Each for $3,383.00
Add to cart
 
Specifications

Specifications

Powder
Dehydrated culture media
1460 Days
Peptone
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit